Learn More
Cayman Chemical Bilastine, Purity: ≥98%, CAS: 202189-78-4, 250 mg
SDP
Catalog No. 502938505
Encompass_Preferred
A histamine H1 receptor antagonist (IC50 = 180 nM); selective for the histamine H1 receptor in a panel of 30 receptors in vitro at 100 µM; prevents microvascular extravasation (ED50 = 185 µg/kg, i.v.), bronchospasm (ED50 = 4.6 µg/kg, i.v.), and systemic anaphylaxis (ED50 = 0.2 µg/kg, p.o.) induced by subcutaneous histamine in guinea pigs; prevents anaphylaxis induced by subcutaneous administration of ovalbumin or DNP in sensitized rats at 7.6 and 6.0 mg/kg, respectively
Catalog No. 50-293-8505 Supplier Cayman Chemical Supplier No. 28375250 MG
A histamine H1 receptor antagonist (IC50 = 180 nM); selective for the histamine H1 receptor in a panel of 30 receptors in vitro at 100 µM; prevents microvascular extravasation (ED50 = 185 µg/kg, i.v.), bronchospasm (ED50 = 4.6 µg/kg, i.v.), and systemic anaphylaxis (ED50 = 0.2 µg/kg, p.o.) induced by subcutaneous histamine in guinea pigs; prevents anaphylaxis induced by subcutaneous administration of ovalbumin or DNP in sensitized rats at 7.6 and 6.0 mg/kg, respectively