Learn More
Cayman Chemical Bilirubin, Purity: ≥95%, CAS: 635-65-4, 1 g
SDP
Catalog No. 502965749
Encompass_Preferred
A yellow breakdown product of heme catabolism, formed when heme is cleaved by heme oxygenase; has key roles as a free radical scavenger with antioxidant and anti-inflammatory actions
Catalog No. 50-296-5749 Supplier Cayman Chemical Supplier No. 171611 G
A yellow breakdown product of heme catabolism, formed when heme is cleaved by heme oxygenase; has key roles as a free radical scavenger with antioxidant and anti-inflammatory actions