Learn More
Cayman Chemical ANTIBODY BIocytIn 10mg
SDP
Catalog No. 502967876
Encompass_Preferred
A conjugate of biotin and lysine used for derivatization or conjugation to proteins and other molecules with a medium-chain length spacer; also used as an anterograde, retrograde, or intracellular neuroanatomical tracer that is fixable with aldehyde-based fixatives
Catalog No. 50-296-7876 Supplier Cayman Chemical Supplier No. 1675110 MG
A conjugate of biotin and lysine used for derivatization or conjugation to proteins and other molecules with a medium-chain length spacer; also used as an anterograde, retrograde, or intracellular neuroanatomical tracer that is fixable with aldehyde-based fixatives