Learn More
Cayman Chemical BKT 140, Purity: ≥95%, CAS: 664334-36-5, 1 mg
SDP
Catalog No. 502990113
Encompass_Preferred
A CXCR4 antagonist (IC50 = 0.91 nM); inhibits HIV-1-induced cytopathogenicity in MT-4 cells (EC50 = 4 nM); reduces CXCL12-induced migration of SUP-T1 leukemia and MDA-MB-231 at 100 nM; induces apoptosis in NB4 leukemia and RPMI-8226 multiple myeloma cancer cells at 8 and 40 µM; reduces pulmonary metastasis in an MDA-MB-231 mouse xenograft model; reduces the incidence of arthritis in a mouse model of collagen-induced arthritis
Catalog No. 50-299-0113 Supplier Cayman Chemical Supplier No. 306891 MG
A CXCR4 antagonist (IC50 = 0.91 nM); inhibits HIV-1-induced cytopathogenicity in MT-4 cells (EC50 = 4 nM); reduces CXCL12-induced migration of SUP-T1 leukemia and MDA-MB-231 at 100 nM; induces apoptosis in NB4 leukemia and RPMI-8226 multiple myeloma cancer cells at 8 and 40 µM; reduces pulmonary metastasis in an MDA-MB-231 mouse xenograft model; reduces the incidence of arthritis in a mouse model of collagen-induced arthritis