Learn More
Cayman Chemical ANTIBODY BKT 140 500ug
SDP
Catalog No. 502990069
Encompass_Preferred
A CXCR4 antagonist (IC50 = 0.91 nM); inhibits HIV-1-induced cytopathogenicity in MT-4 cells (EC50 = 4 nM); reduces CXCL12-induced migration of SUP-T1 leukemia and MDA-MB-231 at 100 nM; induces apoptosis in NB4 leukemia and RPMI-8226 multiple myeloma cancer cells at 8 and 40 µM; reduces pulmonary metastasis in an MDA-MB-231 mouse xenograft model; reduces the incidence of arthritis in a mouse model of collagen-induced arthritis
Catalog No. 50-299-0069 Supplier Cayman Chemical Supplier No. 30689500 UG
A CXCR4 antagonist (IC50 = 0.91 nM); inhibits HIV-1-induced cytopathogenicity in MT-4 cells (EC50 = 4 nM); reduces CXCL12-induced migration of SUP-T1 leukemia and MDA-MB-231 at 100 nM; induces apoptosis in NB4 leukemia and RPMI-8226 multiple myeloma cancer cells at 8 and 40 µM; reduces pulmonary metastasis in an MDA-MB-231 mouse xenograft model; reduces the incidence of arthritis in a mouse model of collagen-induced arthritis