Learn More
Enzo Life Sciences Antibody Blocker/Diluent (100ml)
SDP
Catalog No. 502013357
Encompass_Preferred
The Antibody Blocker/Diluent can be used to block nonspecific staining due to antibodies during immunostaining. It can also be used as a diluent for the POLYVIEW PLUS detection reagents or as a primary antibody diluent. This product is intended for immunohistochemistry and in situ hybridization with formalin-fixed parafiin-embedded tissue sections and frozen sections. The blocker/diluent may also be used with blood smears, cytosmears, and cell preparations. Long Term Storage: +4°C. Applications: IHC ISH (in situ hybridization).
Catalog No. 50-201-3357 Supplier Enzo Life Sciences Supplier No. ENZACC1080100
The Antibody Blocker/Diluent can be used to block nonspecific staining due to antibodies during immunostaining. It can also be used as a diluent for the POLYVIEW PLUS detection reagents or as a primary antibody diluent. This product is intended for immunohistochemistry and in situ hybridization with formalin-fixed parafiin-embedded tissue sections and frozen sections. The blocker/diluent may also be used with blood smears, cytosmears, and cell preparations. Long Term Storage: +4°C. Applications: IHC ISH (in situ hybridization).