Learn More
Cayman Chemical BLT-1, Purity: ≥98%, CAS: 321673-30-7, 25 mg
SDP
Catalog No. 502913144
Encompass_Preferred
An inhibitor of SR-BI; It inhibits uptake of [3H]cholesteryl ester-HDL in ldlA[mSR-BI] cells (IC50 = 0.11 µM; inhibits the efflux of cholesterol from ldlA[mSR-BI] cells to HDL (IC50 = 0.15 µM); induces developmental defects in zebrafish embryos at 8 µM
Catalog No. 50-291-3144 Supplier Cayman Chemical Supplier No. 3498925 MG
An inhibitor of SR-BI; It inhibits uptake of [3H]cholesteryl ester-HDL in ldlA[mSR-BI] cells (IC50 = 0.11 µM; inhibits the efflux of cholesterol from ldlA[mSR-BI] cells to HDL (IC50 = 0.15 µM); induces developmental defects in zebrafish embryos at 8 µM