Learn More
Cayman Chemical BLU-782, Purity: ≥95%, CAS: 2141955-96-4, 5 mg
SDP
Catalog No. 502913527
Encompass_Preferred
An ALK2R206H inhibitor; binds to ALK2R206H (IC50 = ≤10 nM); reduces phosphorylation of SMAD1 at Ser463/465 in HEK293 cells expressing ALK2R206H (IC50 = ≤100 nM)
Catalog No. 50-291-3527 Supplier Cayman Chemical Supplier No. 354905 MG
An ALK2R206H inhibitor; binds to ALK2R206H (IC50 = ≤10 nM); reduces phosphorylation of SMAD1 at Ser463/465 in HEK293 cells expressing ALK2R206H (IC50 = ≤100 nM)