Learn More
Cayman Chemical BMS 753, Purity: ≥98%, CAS: 215307-86-1, 5 mg
SDP
Catalog No. 502989199
Encompass_Preferred
A synthetic retinoid and RARα agonist (Ki = 2 nM); selective for RARα over RARγ at 1 μM; induces morphological differentiation of P19 embryonal carcinoma cells at 10 and 100 nM, as well as F9 embryonal carcinoma cells at 100 nM in combination with BMS649
Catalog No. 50-298-9199 Supplier Cayman Chemical Supplier No. 294935 MG
A synthetic retinoid and RARα agonist (Ki = 2 nM); selective for RARα over RARγ at 1 μM; induces morphological differentiation of P19 embryonal carcinoma cells at 10 and 100 nM, as well as F9 embryonal carcinoma cells at 100 nM in combination with BMS649