Learn More
Cayman Chemical BMS 986158, Purity: ≥98%, CAS: 1800340-40-2, 25 mg
SDP
Catalog No. 502938865
Encompass_Preferred
A BET bromodomain inhibitor; decreases c-Myc expression and induces cancer cell death in c-Myc-driven cancer cell lines in vitro; inhibits tumor growth in triple-negative breast, colorectal, and lung cancer patient-derived mouse xenograft models
Catalog No. 50-293-8865 Supplier Cayman Chemical Supplier No. 2778925 MG
A BET bromodomain inhibitor; decreases c-Myc expression and induces cancer cell death in c-Myc-driven cancer cell lines in vitro; inhibits tumor growth in triple-negative breast, colorectal, and lung cancer patient-derived mouse xenograft models