Learn More
Cayman Chemical Bourgeonal, Purity: ≥95%, CAS: 18127-01-0, 25 mg
SDP
Catalog No. 502988465
Encompass_Preferred
An hOR17-4 agonist; increases cytosolic calcium levels in human spermatozoa at 5 μM; acts as a chemoattractant and increases swimming speed in sperm chemotaxis assays in a concentration-dependent manner
Catalog No. 50-298-8465 Supplier Cayman Chemical Supplier No. 2996525 MG
An hOR17-4 agonist; increases cytosolic calcium levels in human spermatozoa at 5 μM; acts as a chemoattractant and increases swimming speed in sperm chemotaxis assays in a concentration-dependent manner