Learn More
Cayman Chemical BP-1-102, Purity: ≥98%, CAS: 1334493-07-0, 25 mg
SDP
Catalog No. 502938051
Encompass_Preferred
An inhibitor of STAT3; binds to STAT3 (Kd = 504 nM) and inhibits STAT3 binding to an IL-6R/gp130 peptide in a fluorescence polarization assay (IC50 = 4.1 µM); decreases viability and proliferation of STAT3-dependent NIH3T3/v-Src, MDA-MB-231, PANC-1, DU145 and A549 cancer cells, but not STAT3-independent NIH3T3, NIH3T3/vRas, TE-71, and A2780S cancer cells at 10-30 µM; reduces tumor growth and inhibits expression of the STAT3-dependent genes encoding c-Myc, survivin, Bcl-xL, cyclin D1, and VEGF in MDA-MB-231 and A549 mouse xenograft models at 1 and 3 mg/kg
Catalog No. 50-293-8051 Supplier Cayman Chemical Supplier No. 2836825 MG
An inhibitor of STAT3; binds to STAT3 (Kd = 504 nM) and inhibits STAT3 binding to an IL-6R/gp130 peptide in a fluorescence polarization assay (IC50 = 4.1 µM); decreases viability and proliferation of STAT3-dependent NIH3T3/v-Src, MDA-MB-231, PANC-1, DU145 and A549 cancer cells, but not STAT3-independent NIH3T3, NIH3T3/vRas, TE-71, and A2780S cancer cells at 10-30 µM; reduces tumor growth and inhibits expression of the STAT3-dependent genes encoding c-Myc, survivin, Bcl-xL, cyclin D1, and VEGF in MDA-MB-231 and A549 mouse xenograft models at 1 and 3 mg/kg