Learn More
Cayman Chemical BPH-1358, Purity: ≥95%, CAS: 5352-53-4, 50 mg
SDP
Catalog No. 502917779
Encompass_Preferred
An inhibitor of UPPS (IC50 = 0.11 µM for the E. coli and S. aureus enzymes) and FPPS (IC50 = 1.8 µM for the human enzyme); active against E. coli and S. aureus (EC50s = 0.3 and 0.29 nM, respectively); increases survival in a mouse model of S. aureus infection at 10 mg/kg
Catalog No. 50-291-7779 Supplier Cayman Chemical Supplier No. 3740250 MG
An inhibitor of UPPS (IC50 = 0.11 µM for the E. coli and S. aureus enzymes) and FPPS (IC50 = 1.8 µM for the human enzyme); active against E. coli and S. aureus (EC50s = 0.3 and 0.29 nM, respectively); increases survival in a mouse model of S. aureus infection at 10 mg/kg