Learn More
Cayman Chemical BQR-695, Purity: ≥98%, CAS: 1513879-21-4, 10 mg
SDP
Catalog No. 502937848
Encompass_Preferred
An antimalarial compound; inhibits recombinant P. vivax and human PI4K (IC50s = 3.5 and 90 nM, respectively); induces schizont-stage arrest and death of P. falciparum (IC50 = ~70 nM)
Catalog No. 50-293-7848 Supplier Cayman Chemical Supplier No. 2748210 MG
An antimalarial compound; inhibits recombinant P. vivax and human PI4K (IC50s = 3.5 and 90 nM, respectively); induces schizont-stage arrest and death of P. falciparum (IC50 = ~70 nM)