Learn More
Cayman Chemical Bromantane, Purity: ≥98%, CAS: 87913-26-6, 25 mg
SDP
Catalog No. 502935986
Encompass_Preferred
An analytical reference standard categorized as a stimulant; has been used to enhance physical performance in athletes; intended for research and forensic applications
Catalog No. 50-293-5986 Supplier Cayman Chemical Supplier No. 2468825 MG
An analytical reference standard categorized as a stimulant; has been used to enhance physical performance in athletes; intended for research and forensic applications