Learn More
Cayman Chemical ANTIBODY BSJ-04-132 5mg
SDP
Catalog No. 502944408
Encompass_Preferred
A PROTAC that drives Cdk4 degradation; selectively induces the degradation of Cdk4 over Cdk6, IKZF1, and IKZF3 in a cereblon-dependent manner in Jurkat cells at 1 µM
Catalog No. 50-294-4408 Supplier Cayman Chemical Supplier No. 412485 MG
A PROTAC that drives Cdk4 degradation; selectively induces the degradation of Cdk4 over Cdk6, IKZF1, and IKZF3 in a cereblon-dependent manner in Jurkat cells at 1 µM