Learn More
Cayman Chemical BSJ-4-116, Purity: ≥95%, CAS: 2519823-34-6, 5 mg
SDP
Catalog No. 502917535
Encompass_Preferred
A PROTAC that drives Cdk12 degradation; induces time- and concentration-dependent degradation of Cdk12 in Jurkat cells; decreases expressions of various genes associated with DNA double-strand break repair in Jurkat cells at 50 nM; inhibits the growth of MOLT-4 leukemia cells in a cereblon-dependent manner
Catalog No. 50-291-7535 Supplier Cayman Chemical Supplier No. 395645 MG
A PROTAC that drives Cdk12 degradation; induces time- and concentration-dependent degradation of Cdk12 in Jurkat cells; decreases expressions of various genes associated with DNA double-strand break repair in Jurkat cells at 50 nM; inhibits the growth of MOLT-4 leukemia cells in a cereblon-dependent manner