Learn More
Cayman Chemical BSP-16, Purity: ≥98%, CAS: 2727249-47-8, 1 mg
SDP
Catalog No. 502918008
Encompass_Preferred
A STING agonist; binds to STING (Kd = 23 µM); induces the production of IFN-β, IL-6, and CXCL10 in THP-1 cells at 10, 25, and 50 µM; induces phosphorylation of IRF3 and TBK1 in THP-1 cells at 50 µM; reduces tumor volume in an MC-38 murine carcinoma model and induces tumor regression in a CT26 murine colorectal carcinoma model at 15 and 30 mg/kg; prevents tumor cell re-challenge-induced tumor formation in a CT26 murine carcinoma model; reduces tumor volume and increases the number of activated CD8+ T cells and NK cells in a 4T1 murine mammary carcinoma model at 20 mg/kg
Catalog No. 50-291-8008 Supplier Cayman Chemical Supplier No. 379921 MG
A STING agonist; binds to STING (Kd = 23 µM); induces the production of IFN-β, IL-6, and CXCL10 in THP-1 cells at 10, 25, and 50 µM; induces phosphorylation of IRF3 and TBK1 in THP-1 cells at 50 µM; reduces tumor volume in an MC-38 murine carcinoma model and induces tumor regression in a CT26 murine colorectal carcinoma model at 15 and 30 mg/kg; prevents tumor cell re-challenge-induced tumor formation in a CT26 murine carcinoma model; reduces tumor volume and increases the number of activated CD8+ T cells and NK cells in a 4T1 murine mammary carcinoma model at 20 mg/kg