Learn More
Cayman Chemical BT-11, Purity: ≥98%, CAS: 1912399-75-7, 5 mg
SDP
Catalog No. 502978290
Encompass_Preferred
A LANCL2 pathway activator; binds to LANCL2 (Kd = 7.7 µM); increases cAMP levels in mouse splenocytes isolated from wild-type, but not Lancl2-/-, mice at 5 and 10 µM; reduces leukocyte infiltration, epithelial erosion, and mucosal thickening in the colon in a mouse model of DSS-induced colitis at 8 mg/kg; reduces histopathological changes in wild-type mice in a model of DSS-induced colitis but not in mice with a T cell-specific conditional knockout of Lancl2
Catalog No. 50-297-8290 Supplier Cayman Chemical Supplier No. 418565 MG
A LANCL2 pathway activator; binds to LANCL2 (Kd = 7.7 µM); increases cAMP levels in mouse splenocytes isolated from wild-type, but not Lancl2-/-, mice at 5 and 10 µM; reduces leukocyte infiltration, epithelial erosion, and mucosal thickening in the colon in a mouse model of DSS-induced colitis at 8 mg/kg; reduces histopathological changes in wild-type mice in a model of DSS-induced colitis but not in mice with a T cell-specific conditional knockout of Lancl2