Learn More
Cayman Chemical BTTAA, Purity: ≥98%, CAS: 1334179-85-9, 10 mg
SDP
Catalog No. 502978410
Encompass_Preferred
A click chemistry Cu(I)-stabilizing ligand and catalyst; displays an increased CuAAC rate in a propargyl alcohol and 3-azido-7-hydroxycoumarin reaction, as well as an increased yield, compared to the click chemistry ligands BTTES, THPTA, and TBTA in a fluorogenic assay; has been used with biotin-azide in CuAAC for the labeling and detection of azide-tagged glycoconjugates in Jurkat cells, zebrafish, and bacteria
Catalog No. 50-297-8410 Supplier Cayman Chemical Supplier No. 4108910 MG
A click chemistry Cu(I)-stabilizing ligand and catalyst; displays an increased CuAAC rate in a propargyl alcohol and 3-azido-7-hydroxycoumarin reaction, as well as an increased yield, compared to the click chemistry ligands BTTES, THPTA, and TBTA in a fluorogenic assay; has been used with biotin-azide in CuAAC for the labeling and detection of azide-tagged glycoconjugates in Jurkat cells, zebrafish, and bacteria