Learn More
Cayman Chemical BTB-1, Purity: ≥98%, CAS: 86030-08-2, 250 mg
SDP
Catalog No. 502941380
Encompass_Preferred
A reversible Kif18A inhibitor (IC50 = 1.69 μM); selective for Kif18A over a panel of seven kinesins at 100 μM; inhibits Kif18A-dependent microtubule motility in a microtubule gliding assay at 100 μM; induces mitotic accumulation of HeLa cells
Catalog No. 50-294-1380 Supplier Cayman Chemical Supplier No. 27219250 MG
A reversible Kif18A inhibitor (IC50 = 1.69 μM); selective for Kif18A over a panel of seven kinesins at 100 μM; inhibits Kif18A-dependent microtubule motility in a microtubule gliding assay at 100 μM; induces mitotic accumulation of HeLa cells