Learn More
Cayman Chemical BTZ043, Purity: ≥98%, CAS: 1161233-85-7, 1 mg
SDP
Catalog No. 502995454
Encompass_Preferred
An antibiotic with antimycobacterial activity; targets mycobacterial cell wall synthesis by inhibiting DprE1; active against various Mycobacterium species in vitro, including M. smegmatis, M. bovis, and M. tuberculosis (MICs = 4, 2, and 1 ng/ml, respectively), as well as the actinobacterium N. brasiliensis (MIC50 = 0.125 μg/ml); active against M. tuberculosis in RAW 264.7 murine macrophages (MIC = M. tuberculosis cells in lung and spleen in a mouse model of chronic tuberculosis infection at 37.5 or 300 mg/kg for four weeks
Catalog No. 50-299-5454 Supplier Cayman Chemical Supplier No. 229301 MG
An antibiotic with antimycobacterial activity; targets mycobacterial cell wall synthesis by inhibiting DprE1; active against various Mycobacterium species in vitro, including M. smegmatis, M. bovis, and M. tuberculosis (MICs = 4, 2, and 1 ng/ml, respectively), as well as the actinobacterium N. brasiliensis (MIC50 = 0.125 μg/ml); active against M. tuberculosis in RAW 264.7 murine macrophages (MIC = M. tuberculosis cells in lung and spleen in a mouse model of chronic tuberculosis infection at 37.5 or 300 mg/kg for four weeks