Learn More
Cayman Chemical Bufexamac, Purity: ≥98%, CAS: 2438-72-4, 100 mg
SDP
Catalog No. 502936316
Encompass_Preferred
An NSAID and a class IIb HDAC inhibitor; inhibits IFN-α production in PBMCs (EC50 = 8.9 µM); induces hyperacetylation of tubulin (IC50 = 2.9 µM); inhibits LTA4 hydrolase and aminopeptidase activity (IC50s = 15.86 and 11.59 µM, respectively); inhibits 5-LO (IC50 = 27 µM); inhibits the production of LTB4 in neutrophils (IC50 = 12.91 µM) and reduces fMLP-induced neutrophil migration at 50 and 100 µM; reduces neutrophil levels, as well as TNF-α and IL-1β protein levels, in BALF in a mouse model of LPS-induced acute lung injury at 100 mg/kg
Catalog No. 50-293-6316 Supplier Cayman Chemical Supplier No. 26068100 MG
An NSAID and a class IIb HDAC inhibitor; inhibits IFN-α production in PBMCs (EC50 = 8.9 µM); induces hyperacetylation of tubulin (IC50 = 2.9 µM); inhibits LTA4 hydrolase and aminopeptidase activity (IC50s = 15.86 and 11.59 µM, respectively); inhibits 5-LO (IC50 = 27 µM); inhibits the production of LTB4 in neutrophils (IC50 = 12.91 µM) and reduces fMLP-induced neutrophil migration at 50 and 100 µM; reduces neutrophil levels, as well as TNF-α and IL-1β protein levels, in BALF in a mouse model of LPS-induced acute lung injury at 100 mg/kg