Learn More
Cayman Chemical ANTIBODY BX-912 10mg
SDP
Catalog No. 502971250
Encompass_Preferred
A potent, ATP-competitive inhibitor of PDK1 (IC50 = 26 nM)
Catalog No. 50-297-1250 Supplier Cayman Chemical Supplier No. 1470810 MG
A potent, ATP-competitive inhibitor of PDK1 (IC50 = 26 nM)