Learn More
Cayman Chemical BZAD-01, Purity: ≥98%, CAS: 305339-41-7, 25 mg
SDP
Catalog No. 502916479
Encompass_Preferred
An antagonist of NR2B subunit-containing NMDA receptors (Ki = 72 nM); selective for NR2B subunit-containing NMDA receptors over those containing NR2A subunits (IC50s = 47 and >10,000 nM, respectively); reduces carrageenan-induced hyperalgesia in rats (ED50 = 5.5 mg/kg); reduces dopaminergic neuron loss in the rat substantia nigra pars compacta and improves measures of ipsilateral curling, ipsilateral head positioning, and beam walking in a rat model of 6-OHDA-induced Parkinson’s disease at 10 mg/kg
Catalog No. 50-291-6479 Supplier Cayman Chemical Supplier No. 3878825 MG
An antagonist of NR2B subunit-containing NMDA receptors (Ki = 72 nM); selective for NR2B subunit-containing NMDA receptors over those containing NR2A subunits (IC50s = 47 and >10,000 nM, respectively); reduces carrageenan-induced hyperalgesia in rats (ED50 = 5.5 mg/kg); reduces dopaminergic neuron loss in the rat substantia nigra pars compacta and improves measures of ipsilateral curling, ipsilateral head positioning, and beam walking in a rat model of 6-OHDA-induced Parkinson’s disease at 10 mg/kg