Learn More
Cayman Chemical C-87, Purity: ≥95%, CAS: 332420-90-3, 25 mg
SDP
Catalog No. 502914024
Encompass_Preferred
An inhibitor of TNF-α; binds to TNF-α; inhibits TNF-α-induced cytotoxicity (IC50 = 8.73 µM) in L-929 cells; reduces liver inflammation and injury, as well as increases survival, in a mouse model of LPS- and GalN-induced acute hepatitis at 12.5 mg/kg
Catalog No. 50-291-4024 Supplier Cayman Chemical Supplier No. 3657825 MG
An inhibitor of TNF-α; binds to TNF-α; inhibits TNF-α-induced cytotoxicity (IC50 = 8.73 µM) in L-929 cells; reduces liver inflammation and injury, as well as increases survival, in a mouse model of LPS- and GalN-induced acute hepatitis at 12.5 mg/kg