Learn More
Cayman Chemical C-DIM8, Purity: ≥95%, CAS: 151358-47-3, 5 mg
SDP
Catalog No. 502989974
Encompass_Preferred
A Nur77 antagonist; inhibits C-DIM5-induced reporter gene expression in PANC-28 cells at 20 µM; reduces expression of BIRC5 in PANC-1 cells at 10, 15, and 20 µM; induces tumor cell apoptosis and reduces tumor volume in an L3.6pl pancreatic carcinoma mouse xenograft model at 30 mg/kg
Catalog No. 50-298-9974 Supplier Cayman Chemical Supplier No. 306945 MG
A Nur77 antagonist; inhibits C-DIM5-induced reporter gene expression in PANC-28 cells at 20 µM; reduces expression of BIRC5 in PANC-1 cells at 10, 15, and 20 µM; induces tumor cell apoptosis and reduces tumor volume in an L3.6pl pancreatic carcinoma mouse xenograft model at 30 mg/kg