Learn More
Cayman Chemical C12-200, Purity: ≥95%, CAS: 1220890-25-4, 50 mg
SDP
Catalog No. 502914047
Encompass_Preferred
A branched-chain ionizable cationic lipidoid; has been used in combination with other lipids in the formation of LNPs for the delivery of mRNA, siRNA, or saRNA; LNPs containing C12-200 and encapsulating an mRNA reporter selectively accumulate in the liver, and to a lesser extent in the spleen and pancreas, in mice; administration of human EPO mRNA or factor VII siRNA in C12-200-containing LNPs increases serum EPO levels and reduces factor VII serum levels, respectively, in mice
Catalog No. 50-291-4047 Supplier Cayman Chemical Supplier No. 3669950 MG
A branched-chain ionizable cationic lipidoid; has been used in combination with other lipids in the formation of LNPs for the delivery of mRNA, siRNA, or saRNA; LNPs containing C12-200 and encapsulating an mRNA reporter selectively accumulate in the liver, and to a lesser extent in the spleen and pancreas, in mice; administration of human EPO mRNA or factor VII siRNA in C12-200-containing LNPs increases serum EPO levels and reduces factor VII serum levels, respectively, in mice