Learn More
Cayman Chemical C3-K2-E14, Purity: ≥98%, CAS: 2933215-86-0, 10 mg
SDP
Catalog No. 502915711
Encompass_Preferred
An ionizable cationic lipid; has been used in the generation of lipid nanoparticles (LNPs) for the delivery of mRNA, siRNA, and lncRNA in vitro and in vivo; LNPs containing C3-K2-E14 and encapsulating CSF1 siRNA reduce the percentage of circulating Ly6Chi monocytes and increase the percentage of circulating Ly6Cint monocytes in mice; LNPs containing C3-K2-E14 and encapsulating GAPLINC reduce LPS-induced NF-κB activation in a SEAP assay using RAW 264.7 reporter cells
Catalog No. 50-291-5711 Supplier Cayman Chemical Supplier No. 3932310 MG
An ionizable cationic lipid; has been used in the generation of lipid nanoparticles (LNPs) for the delivery of mRNA, siRNA, and lncRNA in vitro and in vivo; LNPs containing C3-K2-E14 and encapsulating CSF1 siRNA reduce the percentage of circulating Ly6Chi monocytes and increase the percentage of circulating Ly6Cint monocytes in mice; LNPs containing C3-K2-E14 and encapsulating GAPLINC reduce LPS-induced NF-κB activation in a SEAP assay using RAW 264.7 reporter cells