Learn More
Cayman Chemical Calcein AM, Purity: ≥90%, CAS: 890090-35-4, 1 mg
SDP
Catalog No. 502973314
Encompass_Preferred
A cell-permeable dye that is cleaved by intracellular esterases, leaving membrane-impermeant calcein, a fluorescent indicator with absorption and emission maxima of 494 and 517 nm, respectively
Catalog No. 50-297-3314 Supplier Cayman Chemical Supplier No. 149481 MG
A cell-permeable dye that is cleaved by intracellular esterases, leaving membrane-impermeant calcein, a fluorescent indicator with absorption and emission maxima of 494 and 517 nm, respectively