Learn More
Cayman Chemical ANTIBODY CalceIn AM 500ug
SDP
Catalog No. 502968517
Encompass_Preferred
A cell-permeable dye that is cleaved by intracellular esterases, leaving membrane-impermeant calcein, a fluorescent indicator with absorption and emission maxima of 494 and 517 nm, respectively
Catalog No. 50-296-8517 Supplier Cayman Chemical Supplier No. 14948500 UG
A cell-permeable dye that is cleaved by intracellular esterases, leaving membrane-impermeant calcein, a fluorescent indicator with absorption and emission maxima of 494 and 517 nm, respectively