Learn More
Cayman Chemical Calpeptin, Purity: ≥90%, CAS: 117591-20-5, 5 mg
SDP
Catalog No. 502970089
Encompass_Preferred
A cell-penetrating peptide inhibitor of calpain (IC50 = 5 nM) which completely abolishes calpain activity in platelets; also a potent inhibitor of cathepsin K (IC50 = 0.11 nM)
Catalog No. 50-297-0089 Supplier Cayman Chemical Supplier No. 145935 MG
A cell-penetrating peptide inhibitor of calpain (IC50 = 5 nM) which completely abolishes calpain activity in platelets; also a potent inhibitor of cathepsin K (IC50 = 0.11 nM)