Learn More
Cayman Chemical CAP 3, Purity: ≥98%, CAS: 2292123-03-4, 5 mg
SDP
Catalog No. 502939263
Encompass_Preferred
A CAP with antibacterial activity; active against the Gram-negative bacteria E. coli, K. pneumoniae, and A. baumanii (MIC99s = 8, 16, and 16 μM, respectively); increases the fluidity of model Gram-negative bacterial membranes and binds to LPS in vitro; reduces the biomass and number of colony-forming units in E. coli biofilms; inhibits E. coli biofilm formation on catheters implanted in mice infected with E. coli at the incision site when applied as a coating on the catheters; reduces bacterial load in E. coli-infected wounds in mice at 40 mg/kg; cytotoxic to A459 cells (IC50 = 56.4 μM) and has hemolytic activity against human red blood cells (HC50 = 48 μM)
Catalog No. 50-293-9263 Supplier Cayman Chemical Supplier No. 275055 MG
A CAP with antibacterial activity; active against the Gram-negative bacteria E. coli, K. pneumoniae, and A. baumanii (MIC99s = 8, 16, and 16 μM, respectively); increases the fluidity of model Gram-negative bacterial membranes and binds to LPS in vitro; reduces the biomass and number of colony-forming units in E. coli biofilms; inhibits E. coli biofilm formation on catheters implanted in mice infected with E. coli at the incision site when applied as a coating on the catheters; reduces bacterial load in E. coli-infected wounds in mice at 40 mg/kg; cytotoxic to A459 cells (IC50 = 56.4 μM) and has hemolytic activity against human red blood cells (HC50 = 48 μM)