Learn More
Cayman Chemical Captan, Purity: ≥98%, CAS: 133-06-2, 100 mg
SDP
Catalog No. 502936903
Encompass_Preferred
A fungicide; reduces radial growth of and starch depletion by A. fumigatus on the leaf surface of A. tenuis at 50 μg/ml; increases the number of micronuclei and chromosomal aberrations in polychromatic erythrocytes in bone marrow in mice from 50-1,000 mg/kg; induces chromosomal aberrations in spermatocytes and sperm head abnormalities in mice; reduces survival of adult male and female alfalfa leafcutting bees from 150-684 and 24.45-48.9 g/L topically and in diet, respectively
Catalog No. 50-293-6903 Supplier Cayman Chemical Supplier No. 24138100 MG
A fungicide; reduces radial growth of and starch depletion by A. fumigatus on the leaf surface of A. tenuis at 50 μg/ml; increases the number of micronuclei and chromosomal aberrations in polychromatic erythrocytes in bone marrow in mice from 50-1,000 mg/kg; induces chromosomal aberrations in spermatocytes and sperm head abnormalities in mice; reduces survival of adult male and female alfalfa leafcutting bees from 150-684 and 24.45-48.9 g/L topically and in diet, respectively