Learn More
Cayman Chemical CASIN, Purity: ≥95%, CAS: 425399-05-9, 5 mg
SDP
Catalog No. 502980092
Encompass_Preferred
A Cdc42 GTPase inhibitor (IC50 = 2 µM for inhibiting actin assembly in PIP2-stimulated actin polymerization assays) that has been used to explore the role of Cdc42 in the division, survival, and differentiation of hematopoietic stem cells
Catalog No. 50-298-0092 Supplier Cayman Chemical Supplier No. 176945 MG
A Cdc42 GTPase inhibitor (IC50 = 2 µM for inhibiting actin assembly in PIP2-stimulated actin polymerization assays) that has been used to explore the role of Cdc42 in the division, survival, and differentiation of hematopoietic stem cells