Learn More
Cayman Chemical CAY10397, Purity: ≥98%, CAS: 78028-01-0, 25 mg
SDP
Catalog No. 502895015
Encompass_Preferred
A selective inhibitor of 15-hydroxy PGDH (IC50 = ~10 µM)
Catalog No. 50-289-5015 Supplier Cayman Chemical Supplier No. 7013025 MG
A selective inhibitor of 15-hydroxy PGDH (IC50 = ~10 µM)