Learn More
Cayman Chemical ANTIBODY CAY10401 100UG
SDP
Catalog No. 502893814
Encompass_Preferred
A selective, potent inhibitor of rat FAAH exhibiting a Ki value of 0.14 nM; approximately 580-fold more potent than oleyl trifluoromethyl ketone when assayed under the same conditions
Catalog No. 50-289-3814 Supplier Cayman Chemical Supplier No. 71652100 UG
A selective, potent inhibitor of rat FAAH exhibiting a Ki value of 0.14 nM; approximately 580-fold more potent than oleyl trifluoromethyl ketone when assayed under the same conditions