Learn More
Cayman Chemical ANTIBODY CAY10662 25ug
SDP
Catalog No. 502954903
Encompass_Preferred
A 1,3 disubstituted urea derivative of 17(R),18(S)-EpETE that is more metabolically robust and reduces the contractility of cardiomyocytes with improved potency (EC50 fold higher than required for the effect on cardiomyocytes, 1-5 μM CAY10662 exerts weak dose-dependent sEH inhibition
Catalog No. 50-295-4903 Supplier Cayman Chemical Supplier No. 1104225 UG
A 1,3 disubstituted urea derivative of 17(R),18(S)-EpETE that is more metabolically robust and reduces the contractility of cardiomyocytes with improved potency (EC50 fold higher than required for the effect on cardiomyocytes, 1-5 μM CAY10662 exerts weak dose-dependent sEH inhibition