Learn More
Cayman Chemical CAY10680, Purity: ≥95%, CAS: 1439488-21-7, 10 mg
SDP
Catalog No. 502971482
Encompass_Preferred
A selective inhibitor of both MAO-B activity (IC50 = 34.9 nM in human) and adenosine A2A receptors (Ki = 39.5 nM in human); designed as a potential dopamine-sparing therapeutic in the treatment of Parkinson’s disease
Catalog No. 50-297-1482 Supplier Cayman Chemical Supplier No. 1561810 MG
A selective inhibitor of both MAO-B activity (IC50 = 34.9 nM in human) and adenosine A2A receptors (Ki = 39.5 nM in human); designed as a potential dopamine-sparing therapeutic in the treatment of Parkinson’s disease