Learn More
Cayman Chemical CAY10722, Purity: ≥98%, CAS: 388086-13-3, 10 mg
SDP
Catalog No. 502986835
Encompass_Preferred
An inhibitor of SIRT3, a class III HDAC (71% inhibition at 200 µM)
Catalog No. 50-298-6835 Supplier Cayman Chemical Supplier No. 2240410 MG
An inhibitor of SIRT3, a class III HDAC (71% inhibition at 200 µM)