Learn More
Cayman Chemical CAY10735, Purity: ≥90%, CAS: 2251753-58-7, 1 mg
SDP
Catalog No. 502934399
Encompass_Preferred
An anticancer compound; inhibits proliferation in a panel of melanoma and breast, pancreatic, and lung cancer cell lines (IC50s = 0.674-11.56 μM); inhibits migration of and EMT in A375 and B16/F10 melanoma cells in vitro in a concentration-dependent manner; reduces the viability of spheroid A375 and B16/F10 cells (IC50s = 3.04 and 1.24 μM, respectively) and increases production of ROS in these cells in a concentration-dependent manner; reduces tumor growth in B16/F10 melanoma and Lewis lung carcinoma mouse models and an A375 mouse xenograft model at 5 mg/kg
Catalog No. 50-293-4399 Supplier Cayman Chemical Supplier No. 267531 MG
An anticancer compound; inhibits proliferation in a panel of melanoma and breast, pancreatic, and lung cancer cell lines (IC50s = 0.674-11.56 μM); inhibits migration of and EMT in A375 and B16/F10 melanoma cells in vitro in a concentration-dependent manner; reduces the viability of spheroid A375 and B16/F10 cells (IC50s = 3.04 and 1.24 μM, respectively) and increases production of ROS in these cells in a concentration-dependent manner; reduces tumor growth in B16/F10 melanoma and Lewis lung carcinoma mouse models and an A375 mouse xenograft model at 5 mg/kg