Learn More
Cayman Chemical CAY10739, Purity: ≥98%, CAS: 2328109-05-1, 10 mg
SDP
Catalog No. 502937674
Encompass_Preferred
A soft-drug agonist of S1P1 (IC50 = 25.12 nM); selective for S1P1 over S1P2, S1P3, and S1P4 (IC50s = 1,000, >10,000, >10,000 nM, respectively); designed to be activated in the skin and rapidly metabolized to avoid systemic effects
Catalog No. 50-293-7674 Supplier Cayman Chemical Supplier No. 2783710 MG
A soft-drug agonist of S1P1 (IC50 = 25.12 nM); selective for S1P1 over S1P2, S1P3, and S1P4 (IC50s = 1,000, >10,000, >10,000 nM, respectively); designed to be activated in the skin and rapidly metabolized to avoid systemic effects