Learn More
Cayman Chemical CAY10746, Purity: ≥95%, CAS: 2247240-76-0, 5 mg
SDP
Catalog No. 502988160
Encompass_Preferred
A ROCK1 and ROCK2 inhibitor (IC50s = 14 and 3 nM, respectively); selective for ROCK1 and ROCK2 over PKA (IC50 = >10,000 nM) and over 387 recombinant human protein kinases in a panel of 394 kinases (IC50s = >10,000 nM for all) but does inhibit LIMK2, Aurora A, Aurora B, PKG1α, and PKG1β (IC50s = 46, 1,072, 1,239, 517, and 660 nM, respectively); inhibits phosphorylation of MYPT1 in SH-SY5Y cells at 0.1-10 µM; inhibits HUVEC migration at 1 µM; protects isolated mouse retinal neurons from apoptosis and oxidative stress induced by high glucose in an in vitro model of diabetic retinopathy; promotes high glucose-induced vessel regression in mouse retinal explants
Catalog No. 50-298-8160 Supplier Cayman Chemical Supplier No. 298145 MG
A ROCK1 and ROCK2 inhibitor (IC50s = 14 and 3 nM, respectively); selective for ROCK1 and ROCK2 over PKA (IC50 = >10,000 nM) and over 387 recombinant human protein kinases in a panel of 394 kinases (IC50s = >10,000 nM for all) but does inhibit LIMK2, Aurora A, Aurora B, PKG1α, and PKG1β (IC50s = 46, 1,072, 1,239, 517, and 660 nM, respectively); inhibits phosphorylation of MYPT1 in SH-SY5Y cells at 0.1-10 µM; inhibits HUVEC migration at 1 µM; protects isolated mouse retinal neurons from apoptosis and oxidative stress induced by high glucose in an in vitro model of diabetic retinopathy; promotes high glucose-induced vessel regression in mouse retinal explants