Learn More
Cayman Chemical CAY10760, Purity: ≥98%, CAS: 391889-85-3, 1 mg
SDP
Catalog No. 502992003
Encompass_Preferred
An inhibitor of the RAD51-BRCA2 protein-protein interaction (EC50 = 19 µM in a cell-free competitive ELISA); decreases homologous recombination by 54% in BxPC-3 pancreatic cancer cells at 20 µM; decreases proliferation of BxPC-3 and Capan-1 cancer cells when used alone or in combination with olaparib
Catalog No. 50-299-2003 Supplier Cayman Chemical Supplier No. 305321 MG
An inhibitor of the RAD51-BRCA2 protein-protein interaction (EC50 = 19 µM in a cell-free competitive ELISA); decreases homologous recombination by 54% in BxPC-3 pancreatic cancer cells at 20 µM; decreases proliferation of BxPC-3 and Capan-1 cancer cells when used alone or in combination with olaparib