Learn More
Cayman Chemical CAY10766, Purity: ≥98%, CAS: 2739990-96-4, 10 mg
SDP
Catalog No. 502990647
Encompass_Preferred
An antiviral compound; inhibits entry of influenza virus-like particles pseutotyped with HA from H5N1 influenza A virus into A549 cells (EC50 = 0.24 µM); reduces H1N1 and H5N1 influenza A viral titers in infected MDCK cells at 1 µM; exhibits synergy with oseltamivir carboxylate in MDCK cells infected with the influenza A reporter PR8-NS1-Gluc virus
Catalog No. 50-299-0647 Supplier Cayman Chemical Supplier No. 3060410 MG
An antiviral compound; inhibits entry of influenza virus-like particles pseutotyped with HA from H5N1 influenza A virus into A549 cells (EC50 = 0.24 µM); reduces H1N1 and H5N1 influenza A viral titers in infected MDCK cells at 1 µM; exhibits synergy with oseltamivir carboxylate in MDCK cells infected with the influenza A reporter PR8-NS1-Gluc virus