Learn More
Cayman Chemical ANTIBODY CAY10770 250ug
SDP
Catalog No. 502991444
Encompass_Preferred
An inhibitor of CYP4Z1 (IC50 = 5.9 µM); selective for CYP4Z1 over CYP4A11, CYP4F2, CYP4F3a, CYP4F3b (IC50s = 187-282 µM) but does inhibit CYP4F8 and CYP4F12 (IC50s = 167 and 91 µM, respectively); inhibits 14,15-EET, 19-HETE, and 14,15-DiHET production by 83, 86, and 80%, respectively, in T47D breast cancer cells expressing CYP4Z1 at 3 µM
Catalog No. 50-299-1444 Supplier Cayman Chemical Supplier No. 31216250 UG
An inhibitor of CYP4Z1 (IC50 = 5.9 µM); selective for CYP4Z1 over CYP4A11, CYP4F2, CYP4F3a, CYP4F3b (IC50s = 187-282 µM) but does inhibit CYP4F8 and CYP4F12 (IC50s = 167 and 91 µM, respectively); inhibits 14,15-EET, 19-HETE, and 14,15-DiHET production by 83, 86, and 80%, respectively, in T47D breast cancer cells expressing CYP4Z1 at 3 µM