Learn More
Cayman Chemical CAY10781, Purity: ≥90%, CAS: 1210360-87-4, 100 mg
SDP
Catalog No. 502948273
Encompass_Preferred
An NRP-1/VEGF-A interaction inhibitor; inhibits the interaction by 43% at 12.5 µM; inhibits VEGF-A-induced phosphorylation of VEGFR2 in CAD cells at 12.5 µM
Catalog No. 50-294-8273 Supplier Cayman Chemical Supplier No. 33778100 MG
An NRP-1/VEGF-A interaction inhibitor; inhibits the interaction by 43% at 12.5 µM; inhibits VEGF-A-induced phosphorylation of VEGFR2 in CAD cells at 12.5 µM