Learn More
Cayman Chemical CAY10789, Purity: ≥98%, CAS: 123226-28-8, 5 mg
SDP
Catalog No. 502913304
Encompass_Preferred
A CysLT1 receptor antagonist (IC50 = 2.1 µM)
Catalog No. 50-291-3304 Supplier Cayman Chemical Supplier No. 353255 MG
A CysLT1 receptor antagonist (IC50 = 2.1 µM)