Learn More
Cayman Chemical CB-7921220, Purity: ≥98%, CAS: 115453-99-1, 10 mg
SDP
Catalog No. 502993786
Encompass_Preferred
An adenylyl cyclase 1 (AC1) and AC6 inhibitor; selective for AC1 and AC6, reducing their activity by 60% while having no effect on AC2 and AC5 in Sf9 membranes stimulated by forskolin (Item No. 11018) at 100 µM
Catalog No. 50-299-3786 Supplier Cayman Chemical Supplier No. 2333210 MG
An adenylyl cyclase 1 (AC1) and AC6 inhibitor; selective for AC1 and AC6, reducing their activity by 60% while having no effect on AC2 and AC5 in Sf9 membranes stimulated by forskolin (Item No. 11018) at 100 µM