Learn More
Cayman Chemical ANTIBODY CBR-5884 5mg
SDP
Catalog No. 502981379
Encompass_Preferred
An inhibitor of PHGDH, inhibiting serine synthesis from 3-PG in cells (IC50 = 33 µM); inhibits the proliferation of cancer cells by blocking de novo serine synthesis
Catalog No. 50-298-1379 Supplier Cayman Chemical Supplier No. 192365 MG
An inhibitor of PHGDH, inhibiting serine synthesis from 3-PG in cells (IC50 = 33 µM); inhibits the proliferation of cancer cells by blocking de novo serine synthesis